Ethyl (trimethylsilyl)acetate ≥93.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Ethyl 2-(trimethylsilyl)acetate, (Trimethylsilyl)acetic acid ethyl ester
T1584-25G T1584-5G
TCIAT1584-25GEA 1725 SEK
TCIAT1584-25G TCIAT1584-5G
Ethyl (trimethylsilyl)acetate ≥93.0%
Ethyl (trimethylsilyl)acetate
Formel: C₇H₁₆O₂Si
MW: 160,29 g/mol
Kokpunkt: 159°C
Flampunkt: 35°C
MDL Number: MFCD00009172
CAS nummer: 4071-88-9
EINECS: 223-783-2
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1de3fd99a8bbe27d08181529e2164c0347b7f196&vid=1de3fd99a8bbe27d08181529e2164c0347b7f196&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %