(R)-(+)-3-Amino-1,2-propanediol ≥98.0%
Leverantör: TCI
|
Fara
|
A1979-1G
A1979-5G
TCIAA1979-1GEA
325
SEK
TCIAA1979-1G
TCIAA1979-5G
(R)-(+)-3-Amino-1,2-propanediol ≥98.0%
(R)-(+)-3-Amino-1,2-propanediol
Formel:
C₃H₉NO₂ Kokpunkt: 163°C |
MDL Number:
MFCD00798261 CAS nummer: 66211-46-9 UN: 3259 ADR: 8,III |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Specific rotation [a]20/D | +2.0 to +4.0 deg (C=5, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...