7,12-Dimethylbenz[a]anthracene ≥98.0%
Leverantör: TCI
Fara
D0677-1G D0677-5G
TCIAD0677-1GEA 1850 SEK
TCIAD0677-1G TCIAD0677-5G
7,12-Dimethylbenz[a]anthracene ≥98.0%
7,12-Dimethylbenz[a]anthracene
Formel: C₂₀H₁₆
Kokpunkt: 440 °C (1013 hPa)
Smältpunkt: 123 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003600
CAS nummer: 57-97-6
EINECS: 200-359-5
ADR: 9,III
Merck Index: 14,03235
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33fa8b3486ef6cb93138d61afd9a3eb28c881506&vid=4bd58e35af65e09c4cae4071acfe2f4a2503cae1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C
Solubility in Toluene almost transparency