(4-Hydroxyphenyl)acetic acid methyl ester ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Methyl 4-hydroxyphenylacetate, Methyl (4-hydroxyphenyl)acetate, Methyl 2-(4-hydroxyphenyl)acetate
H0856-25G
H0856-500G
TCIAH0856-25GEA
525
SEK
TCIAH0856-25G
TCIAH0856-500G
(4-Hydroxyphenyl)acetic acid methyl ester ≥98.0%
(4-Hydroxyphenyl)acetic acid methyl ester
Formel:
C₉H₁₀O₃ MW: 166,18 g/mol Kokpunkt: 310°C |
MDL Number:
MFCD00002387 CAS nummer: 14199-15-6 EINECS: 238-050-2 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 58.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...