9-(Chloromethyl)anthracene ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
C1046-1G C1046-25G C1046-5G
TCIAC1046-1GEA 415 SEK
TCIAC1046-1G TCIAC1046-25G TCIAC1046-5G
9-(Chloromethyl)anthracene ≥98.0% (by GC, titration analysis)
9-(Chloromethyl)anthracene
Formel: C₁₅H₁₁Cl
MW: 226,71 g/mol
Kokpunkt: 371 °C (1013 hPa)
Smältpunkt: 142 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001263
CAS nummer: 24463-19-2
EINECS: 246-274-7
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405a3b530d39580e076a0e315b6cda283cfc3999&vid=eb26c843d4b935ae97b5e5ac3d64d8022a77adb3&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 135.0 to 141.0 deg-C
Solubility in Toluene very faint turbidity