Antracen ≥97.0%
Leverantör: TCI
Varning
A0495-25G A0495-500G A0495-100G
TCIAA0495-25GEA 430 SEK
TCIAA0495-25G TCIAA0495-500G TCIAA0495-100G
Antracen ≥97.0%
Antracen
Formel: C₁₄H₁₀
Kokpunkt: 342°C
Smältpunkt: 111…112 °C
Densitet: 1,24 g/cm³ (20 °C)
Flampunkt: (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00001240
CAS nummer: 120-12-7
EINECS: 204-371-1
UN: 3077
ADR: 9,III
Merck Index: 14,00682
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=656ea8966b19ad2eb0682e50ed47045a04da2538&vid=51b26483d28a8e3f302c2f2902892b09ae0a76aa&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 216.0 to 220.0 deg-C
Solubility in hot Toluene almost transparency