4-Octyne ≥99.0%

Leverantör: TCI
Fara

O0127-5ML O0127-25ML
TCIAO0127-5MLEA 430 SEK
TCIAO0127-5ML TCIAO0127-25ML
4-Octyne ≥99.0%
4-Octyne
Formel: C₈H₁₄
Kokpunkt: 131°C
Smältpunkt: –103 °C
Densitet: 0,7509 g/cm³ (20 °C)
Flampunkt: 29°C
Storage Temperature: Kylskåp
MDL Number: MFCD00009471
CAS nummer: 1942-45-6
EINECS: 217-730-2
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35444090b0c74d4745e11a648088b504014fb131&vid=35444090b0c74d4745e11a648088b504014fb131&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR