2-Hexyne ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
H0429-25ML
H0429-5ML
TCIAH0429-25MLEA
1250
SEK
TCIAH0429-25ML
TCIAH0429-5ML
2-Hexyne ≥98.0%
2-Hexyne
Formel:
C₆H₁₀ MW: 82,15 g/mol Kokpunkt: 84°C Smältpunkt: –91 °C Densitet: 0,7315 g/cm³ (20 °C) Flampunkt: –10°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00009470 CAS nummer: 764-35-2 EINECS: 212-117-6 UN: 3295 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...