1-Hexyne ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
H0140-25ML
H0140-250ML
TCIAH0140-25MLEA
365
SEK
TCIAH0140-25ML
TCIAH0140-250ML
1-Hexyne ≥97.0%
1-Hexyne
Formel:
C₆H₁₀ MW: 82,15 g/mol Kokpunkt: 71°C Smältpunkt: –132 °C Densitet: 0,7147 g/cm³ (20 °C) Flampunkt: –21°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00009504 CAS nummer: 693-02-7 EINECS: 211-736-9 UN: 3295 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...