2-Methyl-1-butene ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
M0174-25ML
M0174-100ML
TCIAM0174-25MLEA
1625
SEK
TCIAM0174-25ML
TCIAM0174-100ML
2-Methyl-1-butene ≥98.0%
2-Methyl-1-butene
Formel:
C₅H₁₀ MW: 70,13 g/mol Kokpunkt: 31°C Smältpunkt: –137,5 °C Densitet: 0,6496 g/cm³ (20 °C) Flampunkt: –20°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00009333 CAS nummer: 563-46-2 EINECS: 209-250-7 UN: 2459 ADR: 3,I |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...