3-Chloro-2-(chloromethyl)propene ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 3-Chloro-2-chloromethyl-1-propene, 3-Chloro-2-(chloromethyl)-1-propene

C1565-25G C1565-5G
TCIAC1565-25GEA 2825 SEK
TCIAC1565-25G TCIAC1565-5G
3-Chloro-2-(chloromethyl)propene ≥98.0%
3-Chloro-2-(chloromethyl)propene
Formel: C₄H₆Cl₂
MW: 125 g/mol
Kokpunkt: 138°C
Smältpunkt: -14°C
Densitet: 1,175 g/cm³ (20 °C)
Flampunkt: 36°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000952
CAS nummer: 1871-57-4
EINECS: 217-489-3
UN: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4ab6c2421a84ac8d160e31e027f498c588c7695&vid=abf8f8752efcf2b66b99ced432de2ff06e567eec&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR