1-Pentene ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: alpha-n-Amylene, Propylethylene

P0316-25ML P0316-500ML P0316-100ML
TCIAP0316-25MLEA 470 SEK
TCIAP0316-25ML TCIAP0316-500ML TCIAP0316-100ML
1-Pentene ≥98.0%
1-Pentene
Formel: C₅H₁₀
MW: 70,13 g/mol
Kokpunkt: 30°C
Smältpunkt: –165 °C
Densitet: 0,641 g/cm³ (20 °C)
Flampunkt: -20 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00003567
CAS nummer: 109-67-1
EINECS: 203-694-5
UN: 1108
ADR: 3,I
Merck Index: 14,07122
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2f594b8ea703e3e1e899b9f4523a4bcff16ae4d&vid=18bc0ccfa7de572862e0d92a7f477b6fc8dd0339&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR