1H,1H,2H-Perfluoro-1-hexene ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
1H,1H,2H-Nonafluorohex-1-ene
N0601-25G
N0601-5G
TCIAN0601-25GEA
1100
SEK
TCIAN0601-25G
TCIAN0601-5G
1H,1H,2H-Perfluoro-1-hexene ≥98.0%
1H,1H,2H-Perfluoro-1-hexene
Formel:
C₆H₃F₉ MW: 246.08 g/mol Kokpunkt: 60°C Densitet: 1.41 Flampunkt: –17°C |
MDL Number:
MFCD00042338 CAS nummer: 19430-93-4 EINECS: 243-053-7 UN: 1993 ADR: 3 II |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light red clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...