Allyl acetate ≥97.0%
Leverantör: TCI
Fara
Synonyms: Acetic acid allyl ester
A0020-25ML A0020-500ML
TCIAA0020-25MLEA 285 SEK
TCIAA0020-25ML TCIAA0020-500ML
Allyl acetate ≥97.0%
Allyl acetate
Formel: C₅H₈O₂
MW: 100.12 g/mol
Kokpunkt: 104°C
Smältpunkt: -96
Flampunkt: 15°C
Storage Temperature: Kylskåp
MDL Number: MFCD00008721
CAS nummer: 591-87-7
EINECS: 209-734-8
UN: 2333
ADR: 3 II
Merck Index: 14,00285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59fa4b542668a791605d720f3258fffb9274ea5e&vid=676136eb8c35625a4ecd98f4bbde0372fe2a3c9c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %