trans-2-Octenal ≥96.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(E)-2-Octenal
O0176-10ML
O0176-25ML
TCIAO0176-10MLEA
670
SEK
TCIAO0176-10ML
TCIAO0176-25ML
trans-2-Octenal ≥96.0%
trans-2-Octenal
Formel:
C₈H₁₄O MW: 126,2 g/mol Kokpunkt: 78°C Flampunkt: 65°C |
MDL Number:
MFCD00007011 CAS nummer: 2548-87-0 EINECS: 219-833-8 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...