trans-2-Nonenal ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (E)-2-Nonenal
N0483-25ML N0483-5ML
TCIAN0483-25MLEA 1325 SEK
TCIAN0483-25ML TCIAN0483-5ML
trans-2-Nonenal ≥95.0%
trans-2-Nonenal
Formel: C₉H₁₆O
MW: 140,23 g/mol
Kokpunkt: 108°C
Densitet: 0,8505 g/cm³ (15 °C)
Flampunkt: 84°C
Storage Temperature: Kylskåp
MDL Number: MFCD00007012
CAS nummer: 18829-56-6
EINECS: 242-609-6
Merck Index: 14,06676
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbcc561830ca71b87124c613dc0cc537e5149c0c&vid=42942869a615b7d36218851684111f81fcdaf641&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %