trans-2-Heptenal ≥95.0%
Leverantör: TCI
|
Fara
|
H0579-25ML
H0579-5ML
TCIAH0579-25MLEA
1600
SEK
TCIAH0579-25ML
TCIAH0579-5ML
trans-2-Heptenal ≥95.0%
trans-2-Heptenal
Formel:
C₇H₁₂O Kokpunkt: 91°C Densitet: 0,8575 g/cm³ (15 °C) Flampunkt: 54°C Storage Temperature: Omgivande |
MDL Number:
MFCD00007010 CAS nummer: 18829-55-5 EINECS: 242-608-0 UN: 1988 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...