cis-4-Heptenal ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: cis-4-Heptenal, Cis-4-heptenal, (Z)-4-Heptenal
H0520-25ML H0520-5ML
TCIAH0520-25MLEA 2675 SEK
TCIAH0520-25ML TCIAH0520-5ML
cis-4-Heptenal ≥95.0%
cis-4-Heptenal
Formel: C₇H₁₂O
MW: 112,17 g/mol
Kokpunkt: 58°C
Densitet: 0,85 g/cm³ (20 °C)
Flampunkt: 44°C
Storage Temperature: Kylskåp
MDL Number: MFCD00014682
CAS nummer: 6728-31-0
EINECS: 229-779-7
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cecd52c2233753b3af142a3c88313dd11815595&vid=1472aa180fd717b5aec0b9c23340b6179f8eae42&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %