4-Bromo-3,3,4,4-tetrafluoro-1-butene ≥98.0%

Leverantör: TCI
Varning

B3222-25G B3222-5G
TCIAB3222-25GEA 2125 SEK
TCIAB3222-25G TCIAB3222-5G
4-Bromo-3,3,4,4-tetrafluoro-1-butene ≥98.0%
4-Bromo-3,3,4,4-tetrafluoro-1-butene
Formel: C₄H₃BrF₄
Kokpunkt: 60°C
Densitet: 1,627 g/cm³ (20 °C)
MDL Number: MFCD00042189
CAS nummer: 18599-22-9
EINECS: 242-440-8
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dd6dd94a1b000b924b2dc8345bb3dcf0b8f2e1f&vid=48fde55598cae802f1a00bce5a50198d441512c5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR