trans-1,4-Dibromo-2-butene ≥98.0%
Leverantör: TCI
Fara
Synonyms: (E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene
D2944-25G D2944-250G
TCIAD2944-25GEA 1025 SEK
TCIAD2944-25G TCIAD2944-250G
trans-1,4-Dibromo-2-butene ≥98.0%
trans-1,4-Dibromo-2-butene
Formel: C₄H₆Br₂
MW: 213.9 g/mol
Kokpunkt: 76°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000249
CAS nummer: 821-06-7
EINECS: 212-472-7
UN: 2923
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4e76fdf29a82a8a97148a15d94ffca2d0db937d&vid=d1e042df6760fc801521d29184bb71e623d2b9f5&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in Methanol almost transparency