Methyl phenylacetate ≥99.0%
Leverantör: TCI
Synonyms: Phenylacetic acid methyll ester
P0125-25ML P0125-500ML
TCIAP0125-25MLEA 230 SEK
TCIAP0125-25ML TCIAP0125-500ML
Methyl phenylacetate ≥99.0%
Methyl phenylacetate
Formel: C₉H₁₀O₂
MW: 150.18 g/mol
Kokpunkt: 215°C
Flampunkt: 89°C
Storage Temperature: Omgivande
MDL Number: MFCD00008453
CAS nummer: 101-41-7
EINECS: 202-940-9
Merck Index: 14,07268
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b5b4125c7192140962c1d39d4ad8af1b0b57916&vid=7393db71febb6d9cb611de384d2e6105f2e4c533&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %