Isopropenyl acetate ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
A0035-25ML
A0035-500ML
TCIAA0035-25MLEA
215
SEK
TCIAA0035-25ML
TCIAA0035-500ML
Isopropenyl acetate ≥98.0%
Isopropenyl acetate
Formel:
C₅H₈O₂ MW: 100,12 g/mol Kokpunkt: 97°C Smältpunkt: –93 °C Densitet: 0,9188 g/cm³ (20 °C) Flampunkt: 16°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008709 CAS nummer: 108-22-5 EINECS: 203-562-7 UN: 2403 ADR: 3,II Merck Index: 14,05203 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...