2,3-Dimethylbutane ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
D0690-25ML
D0690-5ML
D0690-500ML
TCIAD0690-25MLEA
510
SEK
TCIAD0690-25ML
TCIAD0690-5ML
TCIAD0690-500ML
2,3-Dimethylbutane ≥98.0%
2,3-Dimethylbutane
Formel:
C₆H₁₄ MW: 86,18 g/mol Kokpunkt: 58°C Smältpunkt: –128,5 °C Densitet: 0,6615 g/cm³ (20 °C) Flampunkt: –29°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008925 CAS nummer: 79-29-8 EINECS: 201-193-6 UN: 2457 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...