2,2-Dimethylpentane ≥99.0%

Leverantör: TCI EUROPE
Fara

D1673-1ML D1673-5ML
TCIAD1673-1MLEA 335 SEK
TCIAD1673-1ML TCIAD1673-5ML
2,2-Dimethylpentane ≥99.0%
2,2-Dimethylpentane
Formel: C₇H₁₆
MW: 100,2 g/mol
Kokpunkt: 79°C
Smältpunkt: –123,7 °C
Densitet: 0,674 g/cm³ (20 °C)
Flampunkt: -9°C
Storage Temperature: Omgivande
MDL Number: MFCD00009404
CAS nummer: 590-35-2
EINECS: 209-680-5
UN: 1206
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70d3f3585345e6c58f83c500f4771053fa5d437f&vid=70d3f3585345e6c58f83c500f4771053fa5d437f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR