tert-Pentylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Fara

Synonyms: tert-Amylamine

A1002-25ML A1002-500ML
TCIAA1002-25MLEA 455 SEK
TCIAA1002-25ML TCIAA1002-500ML
tert-Pentylamine ≥98.0% (by GC, titration analysis)
tert-Pentylamine
Formel: C₅H₁₃N
MW: 87.16 g/mol
Kokpunkt: 77°C
Smältpunkt: -105
Flampunkt: –1°C
MDL Number: MFCD00008056
CAS nummer: 594-39-8
EINECS: 209-839-9
UN: 1106
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af9357b50ccd1d59bd54d9a4a890a06736f8714e&vid=6ade2d18e91e9cd4114563577f6b86648ace34de&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR