2,6-Dimethyl-3,5-heptanedione ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
D2413-25G
D2413-5G
TCIAD2413-25GEA
3100
SEK
TCIAD2413-25G
TCIAD2413-5G
2,6-Dimethyl-3,5-heptanedione ≥97.0%
2,6-Dimethyl-3,5-heptanedione
Formel:
C₉H₁₆O₂ MW: 156,22 g/mol Kokpunkt: 80°C Densitet: 1,4586 g/cm³ (20 °C) |
MDL Number:
MFCD00015040 CAS nummer: 18362-64-6 EINECS: 242-234-8 UN: 1224 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...