(±)-2-Methylvaleraldehyde ≥95.0%
Leverantör: TCI
|
Fara
|
Synonyms:
2-Methylpentanal, 2-Methylvaleraldehyde
M0596-25ML
M0596-500ML
TCIAM0596-25MLEA
215
SEK
TCIAM0596-25ML
TCIAM0596-500ML
(±)-2-Methylvaleraldehyde ≥95.0%
(±)-2-Methylvaleraldehyde
Formel:
C₆H₁₂O Kokpunkt: 119 °C (1013 hPa) Smältpunkt: –100 °C Densitet: 0,808 g/cm³ (20 °C) Flampunkt: 20°C |
MDL Number:
MFCD00006986 CAS nummer: 123-15-9 EINECS: 204-605-2 UN: 2367 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...