2-Ethylhexanal ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
E0125-25ML
E0125-500ML
TCIAE0125-25MLEA
215
SEK
TCIAE0125-25ML
TCIAE0125-500ML
2-Ethylhexanal ≥95.0%
2-Ethylhexanal
Formel:
C₈H₁₆O MW: 128,21 g/mol Kokpunkt: 163°C Smältpunkt: -85°C Densitet: 0,82 g/cm³ (20 °C) Flampunkt: 46°C Storage Temperature: Omgivande |
MDL Number:
MFCD00006987 CAS nummer: 123-05-7 EINECS: 204-596-5 UN: 1191 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Acid Value | max. 8.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...