2-Ethylbutyraldehyde ≥98.0%
Leverantör: TCI
|
Fara
|
E0069-25ML
E0069-500ML
E0069-100ML
TCIAE0069-25MLEA
270
SEK
TCIAE0069-25ML
TCIAE0069-500ML
TCIAE0069-100ML
2-Ethylbutyraldehyde ≥98.0%
2-Ethylbutyraldehyde
Formel:
C₆H₁₂O Kokpunkt: 117°C Smältpunkt: –89 °C Densitet: 0,8306 g/cm³ (0 °C) Flampunkt: 21°C Storage Temperature: Omgivande |
MDL Number:
MFCD00006985 CAS nummer: 97-96-1 EINECS: 202-623-5 UN: 1178 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...