1-Bromo-3-chloro-2-methylpropane ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
B1859-25G
B1859-500G
TCIAB1859-25GEA
215
SEK
TCIAB1859-25G
TCIAB1859-500G
1-Bromo-3-chloro-2-methylpropane ≥98.0%
1-Bromo-3-chloro-2-methylpropane
Formel:
C₄H₈BrCl MW: 171,46 g/mol Kokpunkt: 154°C Densitet: 1,4839 g/cm³ (20 °C) Flampunkt: ≥ 110 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00000955 CAS nummer: 6974-77-2 EINECS: 230-224-6 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...