n-Oktan ≥97.0%

Leverantör: TCI
Fara

O0022-25ML O0022-500ML O0022-100ML
TCIAO0022-25MLEA 215 SEK
TCIAO0022-25ML TCIAO0022-500ML TCIAO0022-100ML
n-Oktan ≥97.0%
n-Oktan
Formel: H₃C(CH₂)₆CH₃
Kokpunkt: 125°C
Smältpunkt: -57°C
Densitet: 0,703 g/cm³ (20 °C)
Flampunkt: 14°C
Storage Temperature: Omgivande
MDL Number: MFCD00009556
CAS nummer: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,II
Merck Index: 14,06749
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20360300e7be4ae7980f2a9664833bfa5ae5fa34&vid=f5ef4e140aecafc40bd39dd984cb713ec16eaafc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR