4'-Phenoxyacetophenone

Leverantör: TCI EUROPE
Varning

Synonyms: p-Phenoxyacetophenone

P1355-25G P1355-5G
TCIAP1355-25GEA 1175 SEK
TCIAP1355-25G TCIAP1355-5G
4'-Phenoxyacetophenone
4'-Phenoxyacetophenone
Formel: C₁₄H₁₂O₂
MW: 212,25 g/mol
Kokpunkt: 325°C
Smältpunkt: 56 °C
MDL Number: MFCD00008744
CAS nummer: 5031-78-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41e92b93ed57f5855218f54028e3345f2d55af76&vid=41e92b93ed57f5855218f54028e3345f2d55af76&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 53.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR