Propargyl amine ≥97.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 3-Aminopropyne, Prop-2-yn-1-amine, Monopropagyl amine, Prop-2-ynylamine
P0911-25ML P0911-5ML
TCIAP0911-25MLEA 1700 SEK
TCIAP0911-25ML TCIAP0911-5ML
Propargyl amine ≥97.0%
Propargyl amine
Formel: C₃H₅N
MW: 55,08 g/mol
Kokpunkt: 84°C
Densitet: 0,867 g/cm³ (16 °C)
Flampunkt: 6°C
Storage Temperature: Kylskåp
MDL Number: MFCD00008198
CAS nummer: 2450-71-7
EINECS: 219-513-8
UN: 3286
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1efbedb74cd0e26464a7e8c2adb22ee46774a940&vid=1efbedb74cd0e26464a7e8c2adb22ee46774a940&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %