1H,1H-Pentadecafluoro-1-octanol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1H,1H-Perfluoro-1-octanol, 1H,1h-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluor-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octanol

P0904-5G P0904-25G
TCIAP0904-5GEA 415 SEK
TCIAP0904-5G TCIAP0904-25G
1H,1H-Pentadecafluoro-1-octanol ≥98.0%
1H,1H-Pentadecafluoro-1-octanol
Formel: C₈H₃F₁₅O
MW: 400,09 g/mol
Kokpunkt: 164°C
MDL Number: MFCD00004675
CAS nummer: 307-30-2
EINECS: 206-197-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b30379712264fe12907cb922908a0e6a5bf781fd&vid=b30379712264fe12907cb922908a0e6a5bf781fd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR