1H,1H-Tridecafluoro-1-heptanol ≥95.0%

Leverantör: TCI
Varning

Synonyms: 1H,1H-Perfluoro-1-heptanol, 2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoro-1-heptanol

T1701-5G T1701-25G
TCIAT1701-5GEA 885 SEK
TCIAT1701-5G TCIAT1701-25G
1H,1H-Tridecafluoro-1-heptanol ≥95.0%
1H,1H-Tridecafluoro-1-heptanol
Formel: C₇H₃F₁₃O
MW: 350,08 g/mol
Kokpunkt: 95°C
MDL Number: MFCD00042433
CAS nummer: 375-82-6
EINECS: 206-796-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc66ffbb0e8e68096f434081f55746c4d0a16b3&vid=cbc66ffbb0e8e68096f434081f55746c4d0a16b3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR