1-Butylimidazole ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
N-Butyl-1H-imidazole, 1-Butyl-1H-imidazole, N-Butylimidazole
B2345-25G
B2345-100G
TCIAB2345-25GEA
415
SEK
TCIAB2345-25G
TCIAB2345-100G
1-Butylimidazole ≥98.0% (by GC, titration analysis)
1-Butylimidazole
Formel:
C₇H₁₂N₂ MW: 124,19 g/mol Kokpunkt: 235°C Flampunkt: 145°C |
MDL Number:
MFCD00042753 CAS nummer: 4316-42-1 EINECS: 224-335-9 UN: 2690 ADR: 6.1,II |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...