1,11-Dodecadiene 99,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: Dodeca-1,11-diene

D0283-25ML D0283-5ML
TCIAD0283-25MLEA 7925 SEK
TCIAD0283-25ML TCIAD0283-5ML
1,11-Dodecadiene 99,0 GC_ANALYSMETOD
1,11-Dodecadiene
Formel: C₁₂H₂₂
MW: 166.31 g/mol
Kokpunkt: 207°C
MDL Number: MFCD00048156
CAS nummer: 5876-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b484c9a909ce24f1323d7059a358b187ed60c28&vid=9b484c9a909ce24f1323d7059a358b187ed60c28&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR