n-Nonadecane 99,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

Synonyms: N-nonadecane

N0282-25G N0282-400G
TCIAN0282-25GEA 590 SEK
TCIAN0282-25G TCIAN0282-400G
n-Nonadecane 99,0 GC_ANALYSMETOD
n-Nonadecane
Formel: H₃C(CH₂)₁₇CH₃
MW: 268.53 g/mol
Kokpunkt: 329.7
Smältpunkt: 32.5
Storage Temperature: Omgivande
MDL Number: MFCD00009012
CAS nummer: 629-92-5
EINECS: 211-116-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=beb2db320bb2a4d902c68309f42979e341972b27&vid=9857190c55df15efb7830be7421bed9f8956fc2e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 99.0 %
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR