n-Hexacosane ≥99.0%
Leverantör: TCI
Synonyms:
N-hexacosane
H0050-25G
H0050-5G
TCIAH0050-25GEA
885
SEK
TCIAH0050-25G
TCIAH0050-5G
n-Hexacosane ≥99.0%
n-Hexacosane
Formel:
H₃C(CH₂)₂₄CH₃ MW: 366.71 g/mol Kokpunkt: 268°C Storage Temperature: Omgivande |
MDL Number:
MFCD00009354 CAS nummer: 630-01-3 EINECS: 211-124-1 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 99.0 % |
Melting point | 55.0 to 60.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...