2-Propanethiol ≥96.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Isopropyl mercaptan, Isopropanethiol, 2-Mercaptopropane
P0489-25ML
P0489-500ML
TCIAP0489-25MLEA
215
SEK
TCIAP0489-25ML
TCIAP0489-500ML
2-Propanethiol ≥96.0%
2-Propanethiol
Formel:
C₃H₈S Kokpunkt: 53°C Smältpunkt: -130°C Densitet: 0,81 g/cm³ (20 °C) Flampunkt: -34°C Storage Temperature: Omgivande |
MDL Number:
MFCD00004863 CAS nummer: 75-33-2 EINECS: 200-861-4 UN: 2402 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...