1-Propanethiol ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
n-Propanethiol, Propyl mercaptan, n-Propyl mercaptan, 1-Mercaptopropane
P0488-25ML
P0488-500ML
TCIAP0488-25MLEA
405
SEK
TCIAP0488-25ML
TCIAP0488-500ML
1-Propanethiol ≥98.0%
1-Propanethiol
Formel:
C₃H₈S MW: 76,16 g/mol Kokpunkt: 67 °C (1013 hPa) Smältpunkt: –113 °C Densitet: 0,82995 g/cm³ (20 °C) Flampunkt: –20°C Storage Temperature: Omgivande |
MDL Number:
MFCD00004900 CAS nummer: 107-03-9 EINECS: 203-455-5 UN: 2402 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...