1-Octanethiol ≥98.0%

Leverantör: TCI
Varning

Synonyms: n-Octyl mercaptan, 1-Mercaptooctane, n-Octanethiol

O0025-25ML O0025-500ML
TCIAO0025-25MLEA 230 SEK
TCIAO0025-25ML TCIAO0025-500ML
1-Octanethiol ≥98.0%
1-Octanethiol
Formel: C₈H₁₈S
MW: 146,3 g/mol
Kokpunkt: 200°C
Smältpunkt: –49 °C
Densitet: 0,84 g/cm³ (20 °C)
Flampunkt: 81°C
Storage Temperature: Omgivande
MDL Number: MFCD00004912
CAS nummer: 111-88-6
EINECS: 203-918-1
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b15ab44789f3cd8ed8f4c2d84d84c57ac375362&vid=9f8c3fb6bae4007f2883d7d0dfe170e2a50b9ce7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR