Etantiol ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Etylmerkaptan

E0036-25ML E0036-500ML
TCIAE0036-25MLEA 230 SEK
TCIAE0036-25ML TCIAE0036-500ML
Etantiol ≥98.0%
Etantiol
Formel: C₂H₆S
MW: 62,14 g/mol
Kokpunkt: 35°C
Smältpunkt: –148 °C
Densitet: 0,846 g/cm³ (20 °C)
Flampunkt: -48°C
Storage Temperature: Omgivande
MDL Number: MFCD00004887
CAS nummer: 75-08-1
EINECS: 200-837-3
UN: 2363
ADR: 3,I
Merck Index: 14,03726
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34882e21bbd31e06f21192f785a2eb6800df39d2&vid=275722fb1925bacd6b41f94be02c1af92005a805&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR