Hexan-1,2,6-triol ≥96.0%

Leverantör: TCI

H0104-25G H0104-500G
TCIAH0104-25GEA 390 SEK
TCIAH0104-25G TCIAH0104-500G
Hexan-1,2,6-triol ≥96.0%
Hexan-1,2,6-triol
Formel: C₆H₁₄O₃
Kokpunkt: 178°C
Smältpunkt: 25…32 °C
Densitet: 1,11 g/cm³ (20 °C)
Flampunkt: 191°C
Storage Temperature: Omgivande
MDL Number: MFCD00002976
CAS nummer: 106-69-4
EINECS: 203-424-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf8104ede056af342e9fc2a823e58cb1b9aeab70&vid=01789b82caca27c52efe3b0e42b1a70d64b65146&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR