1,9-Nonanediol ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 1,9-Dihydroxy nonane, Nonane-1,9-diol
N0287-25G N0287-500G N0287-100G
TCIAN0287-25GEA 335 SEK
TCIAN0287-25G TCIAN0287-500G TCIAN0287-100G
1,9-Nonanediol ≥98.0%
1,9-Nonanediol
Formel: C₉H₂₀O₂
MW: 160,26 g/mol
Kokpunkt: 171°C
Smältpunkt: 45…46 °C
Densitet: 0,918 g/cm³ (50 °C)
Flampunkt: 162 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002991
CAS nummer: 3937-56-2
EINECS: 223-517-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29a941b2b4969fcf792ab6a94acfda04da9a713d&vid=72b4d358709ee1725d04bbca9848d0bb13751fe7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 49.0 deg-C
Solubility in Methanol almost transparency