1,8-Octanediol ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
1,8-Dihydroxy octane
O0024-25G
O0024-500G
O0024-100G
TCIAO0024-25GEA
550
SEK
TCIAO0024-25G
TCIAO0024-500G
TCIAO0024-100G
1,8-Octanediol ≥99.0%
1,8-Octanediol
Formel:
C₈H₁₈O₂ MW: 146,23 g/mol Kokpunkt: 166°C Smältpunkt: 58…61 °C Densitet: 0,941 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00002989 CAS nummer: 629-41-4 EINECS: 211-090-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 59.0 to 62.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...