4-Heptanol ≥97.0%

Leverantör: TCI
Varning

Synonyms: 4-Heptyl alcohol, Dipropylcarbinol

H0036-25ML H0036-100ML
TCIAH0036-25MLEA 605 SEK
TCIAH0036-25ML TCIAH0036-100ML
4-Heptanol ≥97.0%
4-Heptanol
Formel: C₇H₁₆O
MW: 116.2 g/mol
Kokpunkt: 155°C
Smältpunkt: –42°C
Flampunkt: 57°C
MDL Number: MFCD00021934
CAS nummer: 589-55-9
EINECS: 209-651-7
UN: 1987
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=793d664e05ae968313e16571b10ac0701655d091&vid=793d664e05ae968313e16571b10ac0701655d091&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR