(±)-2-Dodecanol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (±)-Methyl decyl carbinol, (±)-Decyl methyl carbinol, 2-Dodecanol, (±)-2-Hydroxy dodecane

D2960-25G D2960-5G
TCIAD2960-25GEA 4325 SEK
TCIAD2960-25G TCIAD2960-5G
(±)-2-Dodecanol ≥98.0%
(±)-2-Dodecanol
Formel: C₁₂H₂₆O
MW: 186,34 g/mol
Kokpunkt: 129°C
Smältpunkt: 19°C
Densitet: 0,831 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004551
CAS nummer: 10203-28-8
EINECS: 233-500-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3dd12dc6c08f7e8654857b65971a325b66ff073&vid=2905d0d8ac59ba6a92b206b6b1b26ba2a9c12234&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR