1-Undecanol ≥99.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane
U0005-25ML U0005-500ML U0005-100ML
TCIAU0005-25MLEA 230 SEK
TCIAU0005-25ML TCIAU0005-500ML TCIAU0005-100ML
1-Undecanol ≥99.0%
1-Undecanol
Formel: C₁₁H₂₄O
MW: 172,31 g/mol
Kokpunkt: 147°C
Smältpunkt: 13…15 °C
Densitet: 0,83 g/cm³ (20 °C)
Flampunkt: 127°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004751
CAS nummer: 112-42-5
EINECS: 203-970-5
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82ac00bb71f5ceeb2c65d027aeaea8493bc7e480&vid=f49ee5fa6242483470ce8743cf780680c9476395&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %