1-Tetradecanol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol

T0084-25ML T0084-500ML
TCIAT0084-25MLEA 215 SEK
TCIAT0084-25ML TCIAT0084-500ML
1-Tetradecanol ≥98.0%
1-Tetradecanol
Formel: H₃C(CH₂)₁₂CH₂OH
MW: 214,39 g/mol
Kokpunkt: 173°C
Smältpunkt: 37…39 °C
Densitet: 0,8 g/cm³ (60 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004757
CAS nummer: 112-72-1
EINECS: 204-000-3
UN: 3077
ADR: 9,III
Merck Index: 14,06335
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca9d9f427792eeb6956a9ca0b14ca336a6e8498b&vid=7c85acc1010a6addb6796947010fa7204a8508a4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to lump
Purity (GC) min. 98.0 %
Melting point 38.0 to 41.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR